Nitrile gloves, TouchNTuff® 92-605
供应商: Ansell
92-605/SCS 92-605/LCS 92-605/MCS
ANSE92-605/SCSEA 0 CNY
ANSE92-605/SCS ANSE92-605/LCS ANSE92-605/MCS
Nitrile gloves, TouchNTuff® 92-605
手套
Disposable nitrile gloves with extended cuff and proven splash protection against hazardous chemicals.
  • Proprietary Ansell material formulation offers enhanced chemical splash protection
  • Soft nitrile provides high levels of comfort
  • Robust design for superior durability
  • Extended cuff for expanded protection over the wrist and forearm
  • Silicone-free design is paint and finish process friendly
Ideal for laboratory researchers, food processing workers, maintenance workers, and others who need long cuff disposable gloves that protect over the wrist and forearm.
Recommended for: Sample taking and processing; laboratory analysis; transferring liquids and solids; meat, fish and food processing; industrial production; maintenance; chemical handling.
Certifications: EN ISO 374-1:2016 Type B (JKPT); EN ISO 374-5:2016; EN 421; Cat. III; AQL 1,5
Food contact approved
Packaging: 100 gloves per dispenser.