4-氨基安替比林, Supelco®

供应商: Merck
Warning

Synonyms: 氨基安替比林, 4-氨基-2,3-二甲基-1-苯基-3-吡唑啉-5-酮

MQ100

06800-100G 06800-25G
SUPL06800-100GEA 0 CNY
SUPL06800-100G SUPL06800-25G
4-氨基安替比林, Supelco®
4-氨基安替比林
分子式: C₁₁H₁₃N₃O
分子量: 203.24 g/mol
溶点: 107…109 °C
存储条件: Ambient
MDL编号: MFCD00003145
CAS号: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02c5022e409d63d25a340f738ca5312ba147a663&vid=8036f341d4d523a49a91d72204c5290fc2863de2&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR