1-(4-硝基苯基)哌嗪 98%

供应商: Thermo Fisher Scientific
Warning

124940050 124940250
ACRO124940050EA 0 CNY
ACRO124940050 ACRO124940250
1-(4-硝基苯基)哌嗪 98%
1-(4-硝基苯基)哌嗪
分子式: C₁₀H₁₃N₃O₂
溶点: 130…134 °C
存储条件: Ambient
MDL编号: MFCD00005961
CAS号: 6269-89-2
EINECS: 228-443-7
ADR: 6.1,III

现在订购

产品技术规格

Appearance Yellow to ochre Powder
Infrared spectrum Conforms
Melting point 130 to 134 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR