1-(2-羟乙基)哌嗪 98.5%

供应商: Thermo Fisher Scientific
Danger

121191000 121195000
ACRO121191000EA 0 CNY
ACRO121191000 ACRO121195000
1-(2-羟乙基)哌嗪 98.5%
1-(2-羟乙基)哌嗪
分子式: C₆H₁₄N₂O
沸点: ∼246 °C (1013 hPa)
密度: 1,06 g/cm³
闪点: 135 °C
存储条件: Ambient
MDL编号: MFCD00005970
CAS号: 103-76-4
EINECS: 203-142-3

现在订购

产品技术规格

Appearance Clear colorless to light yellow Viscous liquid (May discolor to yellow on storage)
Infrared spectrum Conforms
Refractive index 1.5055 to 1.5075 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR