3,4,5,6-四氯-1,2-苯醌 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 邻四氯苯醌

137990050 137990250
ACRO137990050EA 0 CNY
ACRO137990050 ACRO137990250
3,4,5,6-四氯-1,2-苯醌 97%
3,4,5,6-四氯-1,2-苯醌
分子式: C₆Cl₄O₂
分子量: 245.88 g/mol
沸点: 312
溶点: 125…129 °C
存储条件: Refrigerator
MDL编号: MFCD00001646
CAS号: 2435-53-2
EINECS: 219-424-4

现在订购

产品技术规格

Appearance Bordeaux Crystalline powder
Infrared spectrum Conforms
Melting point 125 to 129 °C
GC ≥96.0 %
Specific optical rotation +12° to +15° (20 °C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR