1,3-二苯基-1,3-丙二酮 98%

供应商: Thermo Fisher Scientific

112600250 112601000
ACRO112600250EA 0 CNY
ACRO112600250 ACRO112601000
1,3-二苯基-1,3-丙二酮 98%
1,3-二苯基-1,3-丙二酮
分子式: (C₆H₅)C(O)CH₂C(O)(C₆H₅)
沸点: 219…221 °C (18 mmHg)
溶点: 75…79 °C
密度: 0,8 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00003085
CAS号: 120-46-7
EINECS: 204-398-9
Merck Index: 13,03033

现在订购

产品技术规格

Appearance Yellow Crystals
Infrared spectrum Conforms
Melting point 75 to 79 °C
Sulfated ash ≤0.1 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR