2-溴-4′-苯基乙酰苯 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 4-苯基苯甲酰甲基溴, ω-溴-4-苯基乙酰苯

146350250 146350050
ACRO146350250EA 0 CNY
ACRO146350250 ACRO146350050
2-溴-4′-苯基乙酰苯 98%
2-溴-4′-苯基乙酰苯
分子式: C₁₄H₁₁BrO
分子量: 275.14 g/mol
沸点: 354
溶点: 123…127 °C
存储条件: Refrigerator
MDL编号: MFCD00000202
CAS号: 135-73-9
EINECS: 205-217-6
UN号: 3261
ADR: 8 II

现在订购

产品技术规格

Appearance Yellow to beige to light brown Powder
Infrared spectrum Conforms
Melting point 123 to 127 °C
GC ≥97.5 %
Specific optical rotation -3.0° to -5.0° (20 °C, 589 nm) (c=2 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR