2-溴-4′-氰基苯乙酮 97%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 4-(2-Bromoacetyl)benzonitrile, 4-(Bromoacetyl)benzonitrile, 4-(2-Bromoacetyl)benzenecarbonitrile, 2-Bromo-4'-cyanoacetophenone, Bromomethyl p-cyanophenyl ketone, 2-Bromo-4′-cyanoacetophenone, 4-Cyanophenacyl bromide, α-Bromo-4-cyanoacetophenone

365360050 365360250
ACRO365360050EA 0 CNY
ACRO365360050 ACRO365360250
2-溴-4′-氰基苯乙酮 97%
2-溴-4′-氰基苯乙酮
分子式: C₉H₆BrNO
分子量: 224.06 g/mol
沸点: 314
溶点: 85…88 °C
MDL编号: MFCD00052931
CAS号: 20099-89-2
UN号: 2928
ADR: 6.1 II

现在订购

产品技术规格

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 96 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR