苯磺酰胺 98+%
供应商: Thermo Fisher Scientific
|
Warning
|
148490050
148495000
ACRO148490050EA
0
CNY
ACRO148490050
ACRO148495000
苯磺酰胺 98+%
苯磺酰胺
分子式:
C₆H₇NO₂S 分子量: 157.19 g/mol 溶点: 151…153 °C 闪点: 250 °C 存储条件: Ambient |
MDL编号:
MFCD00007930 CAS号: 98-10-2 EINECS: 202-637-1 |
产品技术规格
Appearance | White to off-white Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 150 to 154 °C |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...