三(2-甲氧基乙氧基)乙烯基硅烷 ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: 乙烯基三(2-甲氧基乙氧基)硅烷

V0048-25ML V0048-500ML
TCIAV0048-25MLEA 0 CNY
TCIAV0048-25ML TCIAV0048-500ML
三(2-甲氧基乙氧基)乙烯基硅烷 ≥96.0% (by GC)
三(2-甲氧基乙氧基)乙烯基硅烷
分子式: C₁₁H₂₄O₆Si
分子量: 280.39 g/mol
沸点: 113°C
闪点: 132°C
MDL编号: MFCD00008500
CAS号: 1067-53-4
EINECS: 213-934-0

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR