乙烯基磺酸 25% ∼2.3 mol/L in water

供应商: TCI
Warning

Synonyms: 乙烯磺酸, 乙烯基磺酸, 乙烯基磺酸钠

V0043-25ML V0043-500ML
TCIAV0043-25MLEA 0 CNY
TCIAV0043-25ML TCIAV0043-500ML
乙烯基磺酸 25% ∼2.3 mol/L in water
乙烯基磺酸
分子式: C₂H₄O₃S
分子量: 130.09 g/mol
MDL编号: MFCD00007520
CAS号: 3039-83-6
EINECS: 221-242-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8814c65b624102535f57d106f04f372da346035&vid=ac392ed19e2d1dd0ee8e29fbc9b07db55a3264e9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Concentration (Bromination Method) 25.0 to 27.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR