1H,1H-Undecafluorohexylamine ≥97.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 1H,1H-Perfluorohexylamine

U0083-1G U0083-5G
TCIAU0083-1GEA 0 CNY
TCIAU0083-1G TCIAU0083-5G
1H,1H-Undecafluorohexylamine ≥97.0% (by GC, titration analysis)
1H,1H-Undecafluorohexylamine
分子式: C₆H₄F₁₁N
分子量: 299.09 g/mol
沸点: 107°C
MDL编号: MFCD09025929
CAS号: 355-34-0
UN号: 2735
ADR: 8 III

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR