Undecafluoropentyl Iodide ≥98.0% (by GC) stabilised

供应商: TCI
Warning

Synonyms: Perfluoropentyl Iodide

Stabilization: Stabilised with copper chip

U0081-25G U0081-5G
TCIAU0081-25GEA 0 CNY
TCIAU0081-25G TCIAU0081-5G
Undecafluoropentyl Iodide ≥98.0% (by GC) stabilised
Undecafluoropentyl Iodide
分子式: C₅F₁₁I
沸点: 94°C
溶点: –50°C
MDL编号: MFCD08461629
CAS号: 638-79-9
EINECS: 211-350-0

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR