Undecafluoroiodocyclohexane ≥98.0% (by GC) stabilised

供应商: TCI

Synonyms: Perfluoroiodocyclohexane

Stabilization: Stabilised with copper chip

U0080-1G U0080-5G
TCIAU0080-1GEA 0 CNY
TCIAU0080-1G TCIAU0080-5G
Undecafluoroiodocyclohexane ≥98.0% (by GC) stabilised
Undecafluoroiodocyclohexane
分子式: C₆F₁₁I
沸点: 112°C
MDL编号: MFCD00156095
CAS号: 355-69-1

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR