(+)-松萝酸 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Danger

Synonyms: D-2,6-二乙酰基-7,9-二羟基-8,9b-二甲基-1,3(2H,9bH)-二苯并呋喃二酮, (+)-Usnic acid

U0023-1G U0023-25G
TCIAU0023-1GEA 0 CNY
TCIAU0023-1G TCIAU0023-25G
(+)-松萝酸 ≥98.0% (by HPLC, titration analysis)
(+)-松萝酸
分子式: C₁₈H₁₆O₇
分子量: 344.32 g/mol
MDL编号: MFCD00016878
CAS号: 7562-61-0
EINECS: 231-456-0
Merck Index: 14,09893

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 203.0 deg-C
Specific rotation [a]20/D +485 to +505 deg (C=0.7, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR