3-硝基-4-羟乙氨基-N,N-二羟乙基苯胺 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
N1,N4,N4-Tris(2-hydroxyethyl)-2-nitro-1,4-phenylenediamine
T2787-25G
T2787-5G
TCIAT2787-25GEA
0
CNY
TCIAT2787-25G
TCIAT2787-5G
3-硝基-4-羟乙氨基-N,N-二羟乙基苯胺 ≥98.0% (by HPLC, titration analysis)
3-硝基-4-羟乙氨基-N,N-二羟乙基苯胺
分子式:
C₁₂H₁₉N₃O₅ 分子量: 285.30 g/mol |
MDL编号:
MFCD00071762 CAS号: 33229-34-4 EINECS: 251-410-3 |
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...