Trimethylpropylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (通过滴定分析)

供应商: TCI
Warning

T2761-25G T2761-5G
TCIAT2761-25GEA 0 CNY
TCIAT2761-25G TCIAT2761-5G
Trimethylpropylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (通过滴定分析)
Trimethylpropylammonium bis(trifluoromethanesulphonyl)imide
分子式: C₈H₁₆F₆N₂O₄S₂
溶点: 19°C
MDL编号: MFCD08458920
CAS号: 268536-05-6

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR