1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Warning

Synonyms: Trimetazidine Dihydrochloride

T2726-25G T2726-5G
TCIAT2726-25GEA 0 CNY
TCIAT2726-25G TCIAT2726-5G
1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride ≥98.0% (by HPLC, total nitrogen)
1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride
分子式: C₁₄H₂₂N₂O₃
分子量: 339.26 g/mol
溶点: 228°C
MDL编号: MFCD00243086
CAS号: 13171-25-0
EINECS: 236-117-0
Merck Index: 14,09705

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Water max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR