2,4-二氟-α-(1H-1,2,4-三唑基)乙酰苯 ≥98.0% (by GC)
供应商: TCI
Synonyms:
1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone
T2722-25G
T2722-5G
TCIAT2722-25GEA
0
CNY
TCIAT2722-25G
TCIAT2722-5G
2,4-二氟-α-(1H-1,2,4-三唑基)乙酰苯 ≥98.0% (by GC)
2,4-二氟-α-(1H-1,2,4-三唑基)乙酰苯
分子式:
C₁₀H₇F₂N₃O 分子量: 223.18 g/mol |
MDL编号:
MFCD02093825 CAS号: 86404-63-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...