2′,3′,5′-三乙酰鸟苷 ≥98.0% (通过HPLC,滴定分析)

供应商: TCI

T2692-1G T2692-5G
TCIAT2692-1GEA 0 CNY
TCIAT2692-1G TCIAT2692-5G
2′,3′,5′-三乙酰鸟苷 ≥98.0% (通过HPLC,滴定分析)
2′,3′,5′-三乙酰鸟苷
分子式: C₁₆H₁₉N₅O₈
存储条件: Freezer
MDL编号: MFCD00057054
CAS号: 6979-94-8
EINECS: 230-242-4

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -20.0 to -27.0 deg (C=0.4, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR