Tributylmethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (通过滴定分析)

供应商: TCI
Warning

T2679-25G T2679-5G
TCIAT2679-25GEA 0 CNY
TCIAT2679-25G TCIAT2679-5G
Tributylmethylammonium bis(trifluoromethanesulphonyl)imide ≥98.0% (通过滴定分析)
Tributylmethylammonium bis(trifluoromethanesulphonyl)imide
分子式: C₁₅H₃₀F₆N₂O₄S₂
MDL编号: MFCD12761447
CAS号: 405514-94-5

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR