2,4,6-Triphenylboroxin ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 2,4,6-Triphenyl-1,3,5,2,4,6-trioxatriborinane, Phenyl boronic acid anhydride

T2640-25G T2640-5G
TCIAT2640-25GEA 0 CNY
TCIAT2640-25G TCIAT2640-5G
2,4,6-Triphenylboroxin ≥98.0% (by HPLC, titration analysis)
2,4,6-Triphenylboroxin
分子式: C₁₈H₁₅B₃O₃
分子量: 311.75 g/mol
MDL编号: MFCD00014587
CAS号: 3262-89-3

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 217.0 to 221.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR