三(4-咔唑-9-基苯基)胺 ≥98.0% (by HPLC, total nitrogen)
供应商: TCI
|
Warning
|
Synonyms:
TCTA
T2616-1G
T2616-5G
TCIAT2616-1GEA
0
CNY
TCIAT2616-1G
TCIAT2616-5G
三(4-咔唑-9-基苯基)胺 ≥98.0% (by HPLC, total nitrogen)
三(4-咔唑-9-基苯基)胺
分子式:
C₅₄H₃₆N₄ 分子量: 740.91 g/mol 溶点: 266°C |
MDL编号:
MFCD03093250 CAS号: 139092-78-7 |
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...