N-(2-巯基丙酰)-甘氨酸 ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
硫普罗宁
T2614-25G
T2614-5G
TCIAT2614-25GEA
0
CNY
TCIAT2614-25G
TCIAT2614-5G
N-(2-巯基丙酰)-甘氨酸 ≥97.0% (by GC, titration analysis)
N-(2-巯基丙酰)-甘氨酸
分子式:
C₅H₉NO₃S 分子量: 163.19 g/mol |
MDL编号:
MFCD00004861 CAS号: 1953-02-2 EINECS: 217-778-4 Merck Index: 14,09453 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 95.0 to 101.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...