2',3',5'-Tri-O-acetyluridine ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: (2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate, Uridine triacetate, Uridine 2',3',5'-triacetate

T2549-25G T2549-5G
TCIAT2549-25GEA 0 CNY
TCIAT2549-25G TCIAT2549-5G
2',3',5'-Tri-O-acetyluridine ≥98.0% (by HPLC, titration analysis)
2',3',5'-Tri-O-acetyluridine
分子式: C₁₅H₁₈N₂O₉
分子量: 370.31 g/mol
MDL编号: MFCD00023795
CAS号: 4105-38-8
EINECS: 223-881-5

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR