2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate ≥98.0% (by titrimetric analysis)
供应商: TCI
Synonyms:
3-Nitro-1H-1,2,4-triazole-1-carboxylic Acid 2-(Trimethylsilyl)ethyl Ester, 3-Nitro-1-[2-(trimethylsilyl)ethoxycarbonyl]-1H-1,2,4-triazole, Teoc-NT, 2-(Trimethylsilyl)ethyl 3-Nitro-1H-1,2,4-triazole-1-carboxylate
T2544-1G
T2544-5G
TCIAT2544-1GEA
0
CNY
TCIAT2544-1G
TCIAT2544-5G
2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate ≥98.0% (by titrimetric analysis)
2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate
分子式:
C₈H₁₄N₄O₄Si 分子量: 258.31 g/mol 存储条件: Freezer |
MDL编号:
MFCD11042686 CAS号: 1001067-09-9 |
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...