1,2,4-Triazole-3-carboxamide ≥97.0% (by HPLC, total nitrogen)

供应商: TCI

Synonyms: 2H-1,2,4-Triazole-3-carboxamide

T2402-25G T2402-5G
TCIAT2402-25GEA 0 CNY
TCIAT2402-25G TCIAT2402-5G
1,2,4-Triazole-3-carboxamide ≥97.0% (by HPLC, total nitrogen)
1,2,4-Triazole-3-carboxamide
分子式: C₃H₄N₄O
溶点: 315°C
MDL编号: MFCD03990481
CAS号: 3641-08-5

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR