L-Tryptophanamide hydrochloride ≥98.0% (通过HPLC,总氮)

供应商: TCI

T2312-1G T2312-5G
TCIAT2312-1GEA 0 CNY
TCIAT2312-1G TCIAT2312-5G
L-Tryptophanamide hydrochloride ≥98.0% (通过HPLC,总氮)
L-Tryptophanamide hydrochloride
分子式: C₁₁H₁₃N₃O
MDL编号: MFCD00054315
CAS号: 5022-65-1

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +21.0 to +28.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR