三甲基硅基环戊二烯,异构体混合物 mixture of isomers ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Trimethylsilylcyclopentadiene (mixture of isomers), (2,4-Cyclopentadien-1-yl)trimethylsilane
T2284-1G
T2284-5G
TCIAT2284-1GEA
0
CNY
TCIAT2284-1G
TCIAT2284-5G
三甲基硅基环戊二烯,异构体混合物 mixture of isomers ≥97.0% (by GC)
三甲基硅基环戊二烯,异构体混合物
分子式:
C₈H₁₄Si 分子量: 138.29 g/mol 沸点: 140°C 溶点: –19°C 闪点: 29°C 存储条件: Freezer |
MDL编号:
MFCD00010666 CAS号: 25134-15-0 UN号: 1993 ADR: 3 III |
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...