Tetramethylpropylcyclopentadiene (mixture of isomers) ≥95.0% (by GC)
供应商: TCI
T2241-1G
T2241-5G
TCIAT2241-1GEA
0
CNY
TCIAT2241-1G
TCIAT2241-5G
Tetramethylpropylcyclopentadiene (mixture of isomers) ≥95.0% (by GC)
Tetramethylpropylcyclopentadiene (mixture of isomers)
|
产品技术规格
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 95.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...