1-三氟乙酰基哌啶 ≥98.0% (by GC)

供应商: TCI

Synonyms: 1-Trifluoroacetyl Piperidine

T2087-25G T2087-5G
TCIAT2087-25GEA 0 CNY
TCIAT2087-25G TCIAT2087-5G
1-三氟乙酰基哌啶 ≥98.0% (by GC)
1-三氟乙酰基哌啶
分子式: C₇H₁₀F₃NO
分子量: 181.16 g/mol
沸点: 77°C
闪点: 76°C
MDL编号: MFCD00466213
CAS号: 340-07-8

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR