1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranose ≥97.0% (by HPLC)

供应商: TCI

Synonyms: 1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranose

T2047-25G T2047-5G
TCIAT2047-25GEA 0 CNY
TCIAT2047-25G TCIAT2047-5G
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranose ≥97.0% (by HPLC)
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranose
分子式: C₂₂H₂₃NO₁₁
分子量: 477.42 g/mol
溶点: 200°C
存储条件: Freezer
MDL编号: MFCD00080781
CAS号: 10022-13-6

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D +66.0 to +70.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR