2,3,4,6-Tetra-O-benzyl-ɑ-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate
供应商: TCI
T1999-200MG
T1999-1G
TCIAT1999-200MGEA
0
CNY
TCIAT1999-200MG
TCIAT1999-1G
2,3,4,6-Tetra-O-benzyl-ɑ-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate
2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate
分子式:
C₅₀H₄₅F₆NO₆S 分子量: 901.96 g/mol 存储条件: Freezer |
MDL编号:
MFCD06797177 CAS号: 468095-63-8 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Elemental analysis (Carbon) | 65.00 to 68.00 % |
Melting point | 120.0 to 125.0 deg-C |
Specific rotation [a]20/D | +60.0 to +65.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...