2,3,4,6-Tetra-O-benzoyl-ɑ-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate ≥96.0% (通过HPLC)
供应商: TCI
T1991-1G
T1991-200MG
TCIAT1991-1GEA
0
CNY
TCIAT1991-1G
TCIAT1991-200MG
2,3,4,6-Tetra-O-benzoyl-ɑ-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate ≥96.0% (通过HPLC)
2,3,4,6-Tetra-O-benzoyl-alpha-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate
分子式:
C₅₀H₃₇F₆NO₁₀S 存储条件: Freezer |
MDL编号:
MFCD06797174 CAS号: 428816-48-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Specific rotation [a]20/D | +77.0 to +82.0 deg (C=1, Chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...