Tris(2-methoxyphenyl)bismuth dichloride ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
Dichlorotris(2-methoxyphenyl)bismuth
T1956-1G
T1956-5G
TCIAT1956-1GEA
0
CNY
TCIAT1956-1G
TCIAT1956-5G
Tris(2-methoxyphenyl)bismuth dichloride ≥98.0% (by titrimetric analysis)
Tris(2-methoxyphenyl)bismuth dichloride
分子式:
C₂₁H₂₁BiCl₂O₃ 分子量: 601.28 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD04038429 CAS号: 121899-81-8 |
产品技术规格
Purity (Chelometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...