2,4,6-Tris(3,4,5-trifluorophenyl)boroxin ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: Tris(3,4,5-trifluorophenyl)cycloboroxane, 3,4,5-Trifluorophenylboronic Anhydride

T1929-1G T1929-5G
TCIAT1929-1GEA 0 CNY
TCIAT1929-1G TCIAT1929-5G
2,4,6-Tris(3,4,5-trifluorophenyl)boroxin ≥98.0% (by titrimetric analysis)
2,4,6-Tris(3,4,5-trifluorophenyl)boroxin
分子式: C₁₈H₆B₃F₉O₃
分子量: 473.66 g/mol
MDL编号: MFCD03844809
CAS号: 223440-94-6

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR