3-(三氟乙酰基)吲哚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
T1809-1G
T1809-5G
TCIAT1809-1GEA
0
CNY
TCIAT1809-1G
TCIAT1809-5G
3-(三氟乙酰基)吲哚 ≥98.0% (by GC)
3-(三氟乙酰基)吲哚
分子式:
C₁₀H₆F₃NO |
MDL编号:
MFCD00182114 CAS号: 14618-45-2 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 211.0 to 216.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...