2,4,6-Triamino-5-nitrosopyrimidine ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

T1705-25G T1705-5G
TCIAT1705-25GEA 0 CNY
TCIAT1705-25G TCIAT1705-5G
2,4,6-Triamino-5-nitrosopyrimidine ≥98.0% (by HPLC, titration analysis)
2,4,6-Triamino-5-nitrosopyrimidine
分子式: C₄H₆N₆O
MDL编号: MFCD00006096
CAS号: 1006-23-1
EINECS: 213-742-7

现在订购

产品技术规格

Appearance Light red to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR