三乙二醇二(对甲苯磺酸酯) ≥97.0% (by HPLC)

供应商: TCI
Warning

Synonyms: 2,2′-(乙烯二氧)二乙基二甲苯磺酸酯, 三乙二醇二甲苯磺酸酯

T1488-10G T1488-25G
TCIAT1488-10GEA 0 CNY
TCIAT1488-10G TCIAT1488-25G
三乙二醇二(对甲苯磺酸酯) ≥97.0% (by HPLC)
三乙二醇二(对甲苯磺酸酯)
分子式: C₂₀H₂₆O₈S₂
分子量: 458.54 g/mol
MDL编号: MFCD00048096
CAS号: 19249-03-7

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR