Ethyl L-(-)-O-tosyllactate ≥98.0% (通过滴定分析)

供应商: TCI

T1241-1G T1241-25G
TCIAT1241-1GEA 0 CNY
TCIAT1241-1G TCIAT1241-25G
Ethyl L-(-)-O-tosyllactate ≥98.0% (通过滴定分析)
Ethyl L-(-)-O-tosyllactate
分子式: C₁₂H₁₆O₅S
MDL编号: MFCD06809725
CAS号: 57057-80-4

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (Ester Value) min. 98.0 %
Specific rotation [a]20/D -35.0 to -38.0 deg (C=5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR