Levamisole hydrochloride ≥99.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: (-)-Tetramisole Hydrochloride, (-)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole Hydrochloride, (S)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride

T1215-10G T1215-25G
TCIAT1215-10GEA 0 CNY
TCIAT1215-10G TCIAT1215-25G
Levamisole hydrochloride ≥99.0% (by titrimetric analysis)
Levamisole hydrochloride
分子式: C₁₁H₁₂N₂S·HCl
分子量: 240.75 g/mol
MDL编号: MFCD00012675
CAS号: 16595-80-5
EINECS: 240-654-6
UN号: 2811
ADR: 6.1 III
Merck Index: 14,05459

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 226.0 to 234.0 deg-C
Specific rotation [a]20/D -122 to -130 deg (C=1, H2O)
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR