22-Tricosenoic acid ≥97.0% (通过滴定分析)

供应商: TCI
Warning

T1211-1G T1211-100MG
TCIAT1211-1GEA 0 CNY
TCIAT1211-1G TCIAT1211-100MG
22-Tricosenoic acid ≥97.0% (通过滴定分析)
22-Tricosenoic acid
分子式: C₂₃H₄₄O₂
MDL编号: MFCD00060118
CAS号: 65119-95-1

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR