Trimethylhexamethylene diisocyanate (2,2,4- and 2,4,4- mixture) ≥97.0% (通过GC)
供应商: TCI
|
Danger
|
T1176-25ML
T1176-500ML
TCIAT1176-25MLEA
0
CNY
TCIAT1176-25ML
TCIAT1176-500ML
Trimethylhexamethylene diisocyanate (2,2,4- and 2,4,4- mixture) ≥97.0% (通过GC)
Trimethylhexamethylene diisocyanate (2,2,4- and 2,4,4- mixture)
分子式:
C₁₁H₁₈N₂O₂ 存储条件: Refrigerator |
MDL编号:
MFCD00040422 CAS号: 28679-16-5 EINECS: 249-151-6 UN号: 2328 ADR: 6.1,III |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...