5,6,7,8-Tetrahydroquinoline ≥96.0% (通过GC,滴定分析)
供应商: TCI
|
Warning
|
T1150-100ML
T1150-10ML
TCIAT1150-100MLEA
0
CNY
TCIAT1150-100ML
TCIAT1150-10ML
5,6,7,8-Tetrahydroquinoline ≥96.0% (通过GC,滴定分析)
5,6,7,8-Tetrahydroquinoline
分子式:
C₉H₁₁N 沸点: 222°C 闪点: 86°C |
MDL编号:
MFCD00006734 CAS号: 10500-57-9 EINECS: 234-030-2 |
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...