Methyl-2-thiophenecarboxylate ≥99.0% (by GC)

供应商: TCI

Synonyms: Methyl thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Methyl Ester

T1139-25G T1139-5G
TCIAT1139-25GEA 0 CNY
TCIAT1139-25G TCIAT1139-5G
Methyl-2-thiophenecarboxylate ≥99.0% (by GC)
Methyl-2-thiophenecarboxylate
分子式: C₆H₆O₂S
分子量: 142.17 g/mol
沸点: 98°C
MDL编号: MFCD00016895
CAS号: 5380-42-7
EINECS: 226-371-0

现在订购

产品技术规格

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR