2-Thiopheneacrylic acid ≥98.0% (通过滴定分析)

供应商: TCI
Warning

T1138-25G T1138-5G
TCIAT1138-25GEA 0 CNY
TCIAT1138-25G TCIAT1138-5G
2-Thiopheneacrylic acid ≥98.0% (通过滴定分析)
2-Thiopheneacrylic acid
分子式: C₇H₆O₂S
MDL编号: MFCD00005455
CAS号: 15690-25-2

现在订购

产品技术规格

Purity (Neutralization titration) min. 98.0 %
Melting point 145.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR