康洁新 98,0 HPLC, N

供应商: TCI
Warning

Synonyms: 三氯均二苯脲, 三氯卡班, TCC

T1015-25G T1015-500G
TCIAT1015-25GEA 0 CNY
TCIAT1015-25G TCIAT1015-500G
康洁新 98,0 HPLC, N
康洁新
分子式: C₁₃H₉Cl₃N₂O
分子量: 315.58 g/mol
MDL编号: MFCD00013254
CAS号: 101-20-2
EINECS: 202-924-1
UN号: 3077
ADR: 9 III
Merck Index: 14,09654

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in hot 2-Propanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR