Tridecylamine (=monotridecylamine) (mixture of branched chain isomers) ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

T0936-25ML T0936-500ML
TCIAT0936-25MLEA 0 CNY
TCIAT0936-25ML TCIAT0936-500ML
Tridecylamine (=monotridecylamine) (mixture of branched chain isomers) ≥98.0% (by titrimetric analysis)
Tridecylamine (=monotridecylamine) (mixture of branched chain isomers)
分子式: C₁₃H₂₉N
分子量: 199.38 g/mol
MDL编号: MFCD00008156
CAS号: 86089-17-0
EINECS: 289-185-9
UN号: 2735
ADR: 8 III

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR