1,3,5-三甲基-2,4,6-三(3,5-二叔丁基-4-羟基苄基)苯 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1,3,5-Trimethyl-2,4,6-tris(3,5-di-tert-butyl-4-hydroxybenzyl)benzene, 4,4',4''-((2,4,6-Trimethylbenzene-1,3,5-triyl)tris(methylene))tris(2,6-di-tert-butylphenol)
T0916-25G
T0916-500G
TCIAT0916-25GEA
0
CNY
TCIAT0916-25G
TCIAT0916-500G
1,3,5-三甲基-2,4,6-三(3,5-二叔丁基-4-羟基苄基)苯 ≥95.0% (by GC)
1,3,5-三甲基-2,4,6-三(3,5-二叔丁基-4-羟基苄基)苯
分子式:
C₅₄H₇₈O₃ 分子量: 775.22 g/mol |
MDL编号:
MFCD00026284 CAS号: 1709-70-2 EINECS: 216-971-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 240.0 to 245.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...