4,4'-异亚丙基双[2-(2,6-二溴苯氧基)乙醇] ≥93.0% (通过GC)
供应商: TCI
|
Warning
|
T0851-25G
T0851-500G
TCIAT0851-25GEA
0
CNY
TCIAT0851-25G
TCIAT0851-500G
4,4'-异亚丙基双[2-(2,6-二溴苯氧基)乙醇] ≥93.0% (通过GC)
4,4'-异亚丙基双[2-(2,6-二溴苯氧基)乙醇]
分子式:
C₁₉H₂₀Br₄O₄ |
MDL编号:
MFCD00059603 CAS号: 4162-45-2 EINECS: 224-005-4 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 93.0 % |
Melting point | 117.0 to 121.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...